Wood's alloy (lipowitz's alloy), sticks, Sigma-Aldrich®

Danger

Synonyms: Wood's metal, Bismuth Lead Cadmium Tin alloy (50:25:12.5:12.5 w%)

M-Clarity™ quality level = MQ200

244104-100G 244104-500G
220046-976EA 122.31 USD
220046-976 220046-977
Wood's alloy (lipowitz's alloy), sticks, Sigma-Aldrich®
Wood's alloy (lipowitz's alloy)
Melting Pt: 75 °C
Density: 9.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00198175
CAS Number: 76093-98-6
UN: 3288
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR