4-Bromophenyl isocyanate ≥99%

Synonyms: 1-Bromo-4-isocyanatobenzene

241563-10G 241563-50G
220047-158EA 92.62 USD
220047-158 220047-159
4-Bromophenyl isocyanate ≥99%
4-Bromophenyl isocyanate
Formula: C₇H₄BrNO
MW: 198.02 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 42 °C
MDL Number: MFCD00002022
CAS Number: 2493-02-9
EINECS: 219-662-9
UN: 2811
ADR: 6.1,II
Merck Index: 13,01415

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR