Cyclohexylamine ≥99%, ReagentPlus®

Danger

Synonyms: Aminocyclohexane

240648-100ML 240648-5ML
220047-092EA 39.85 USD
220047-092 220047-093
Cyclohexylamine ≥99%, ReagentPlus®
Cyclohexylamine
Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 134.5 °C (1013 hPa)
Melting Pt: -18 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00001486
CAS Number: 108-91-8
EINECS: 203-629-0
UN: 2357
ADR: 8,II
Merck Index: 13,02758

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR