1,4-Butanediol ≥99%, ReagentPlus®

Warning

Synonyms: 1,4-Dihydroxy butane, BDO, Tetramethylene glycol, 1,4-Butylene glycol

240559-100G 240559-2G
220047-089EA 76.92 USD
220047-089 220047-090
1,4-Butanediol ≥99%, ReagentPlus®
1,4-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 19.5 - 20 °C
Density: 1.017 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002968
CAS Number: 110-63-4
EINECS: 203-786-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR