Valeric acid ≥99%

Danger

Synonyms: Pentanoic acid

240370-5ML 240370-100ML
220047-074EA 33.38 USD
220047-074 220047-073
Valeric acid ≥99%
Valeric acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 186 °C (1013 hPa)
Melting Pt: -32 °C
Density: 1.11 g/cm³ (20 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00004413
CAS Number: 109-52-4
EINECS: 203-677-2
UN: 3265
ADR: 8,III
Merck Index: 12,10042

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR