3-Amino-2-cyclohexen-1-one ≥99%

239372-5G 239372-25G
220046-255EA 211.15 USD
220046-255 220046-254
3-Amino-2-cyclohexen-1-one ≥99%
3-Amino-2-cyclohexen-1-one
Formula: C₆H₉NO
MW: 111.14 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: 60 °C
MDL Number: MFCD00013783
CAS Number: 5220-49-5
EINECS: 226-014-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR