Aluminium nitrate nonahydrate ≥98% ACS

Warning

Synonyms: Aluminum nitrate nonahydrate

237973-100G 237973-2.5KG
220046-226EA 58.4 USD
220046-226 220046-627 220046-225
Aluminium nitrate nonahydrate ≥98% ACS
Aluminium nitrate nonahydrate
Formula: Al(NO₃)₃·9H₂O
MW: 375.13 g/mol
Melting Pt: 73 °C
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149132
CAS Number: 7784-27-2
EINECS: 236-751-8
UN: 1438
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR