(R)-(-)-2-Butanol ≥99%

Danger

Synonyms: (R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol

236691-10G 236691-1G
220046-351EA 816.31 USD
220046-351 220046-352
(R)-(-)-2-Butanol ≥99%
(R)-(-)-2-Butanol
Formula: C₄H₁₀O
MW: 74.12 g/mol
Boiling Pt: 99 °C (1013 hPa)
Melting Pt: -114 °C
Density: 0.807 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064280
CAS Number: 14898-79-4
EINECS: 238-967-8
UN: 1120
ADR: 3,III
Merck Index: 13,01540

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR