Sodium hydrogen sulfate monohydrate ≥99%, ReagentPlus®

Danger

233714-1KG 233714-500G 233714-100G
220043-764EA 117.54 USD
220043-764 220043-765 220043-763
Sodium hydrogen sulfate monohydrate ≥99%, ReagentPlus®
Sodium hydrogen sulphate monohydrate
Formula: NaHSO₄·H₂O
Melting Pt: 183 °C
Density: 2.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149210
CAS Number: 10034-88-5
EINECS: 231-665-7
UN: 3260
ADR: 8,III
Merck Index: 13,08658

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR