Chloramine-T (sodium salt) trihydrate, purum p.a.

Danger

Synonyms: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt

23270-250G 23270-1KG 23270-50G
220046-107EA 36.54 USD
220046-107 220046-106 220046-108
Chloramine-T (sodium salt) trihydrate, purum p.a.
Chloramine-T (sodium salt) trihydrate
Formula: CH₃C₆H₄SO₂NClNa·3H₂O
MW: 281.69 g/mol
Melting Pt: 167…170 °C
Density: 1.43 g/cm³ (20 °C)
Flash Pt: 192 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149066
CAS Number: 7080-50-4
EINECS: 204-854-7
UN: 3263
ADR: 8,III
Merck Index: 13,02085

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR