Cobalt(II) chloride

Danger

232696-500G 232696-5G 232696-100G
220046-104EA 363.08 USD
220046-104 220046-105 220046-103
Cobalt(II) chloride
Cobalt(II) chloride
Formula: Cl₂Co
Boiling Pt: 1049 °C (1013 hPa)
Melting Pt: 735 °C
Density: 1.6795 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00010938
CAS Number: 7646-79-9
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 12,02498

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR