Chromium(III) chloride hexahydrate

Warning

Synonyms: Chromium trichloride hexahydrate, Chromic chloride hexahydrate

230723-500G 230723-100G
220045-943EA 55.58 USD
220045-943 220045-942
Chromium(III) chloride hexahydrate
Chromium(III) chloride hexahydrate
Formula: CrCl₃·6H₂O
MW: 266.45 g/mol
Melting Pt: 85 °C
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149660
CAS Number: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8,III
Merck Index: 13,02242

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR