Boron tribromide ≥99.99%

Danger

230367-100G 230367-25G 230367-5G
220045-064EA 217.54 USD
220045-064 220045-065 220045-066
Boron tribromide ≥99.99%
Boron tribromide
Formula: BBr₃
Boiling Pt: 91.2 °C (1013 hPa)
Melting Pt: –46 °C
Density: 2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011312
CAS Number: 10294-33-4
EINECS: 233-657-9
UN: 2692
ADR: 8,I
Merck Index: 13,01337

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR