di-Sodium L(+)-tartrate dihydrate ≥99% ACS

Synonyms: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate

228729-500G 228729-100G 228729-5G
220045-799EA 92.31 USD
220045-799 220045-798 220045-800
di-Sodium L(+)-tartrate dihydrate ≥99% ACS
di-Sodium L(+)-tartrate dihydrate
Formula: NaOOC(CHOH)₂COONa·2H₂O
MW: 230.08 g/mol
Melting Pt: 57 °C
Density: 1.818 g/cm³ (20 °C)
MDL Number: MFCD00150035
CAS Number: 6106-24-7
EINECS: 212-773-3
Merck Index: 12,08833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR