Titanium(IV) oxide rutile

Synonyms: Pigment White 6, Rutile, Titanium dioxide rutile

224227-500G 224227-100G
220045-211EA 492.31 USD
220045-211 220045-210
Titanium(IV) oxide rutile
Titanium(IV) oxide rutile
Formula: O₂Ti
MW: 79.88 g/mol
Melting Pt: 1855 °C
Density: 4.197 g/cm³ (25 °C)
MDL Number: MFCD00011269
CAS Number: 1317-80-2
EINECS: 215-282-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR