Bismuth(III) oxide ≥99.9%, ReagentPlus®

Warning

Synonyms: Bismuth Trioxide, Dibismuth trioxide

223891-100G 223891-500G
220045-193EA 113.23 USD
220045-193 220045-194
Bismuth(III) oxide ≥99.9%, ReagentPlus®
Bismuth(III) oxide
Formula: BiO₃
MW: 465.96 g/mol
Boiling Pt: 1890 °C (1013 hPa)
Melting Pt: 825 °C
Density: 8.9 g/cm³ (20 °C)
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR