Zinc iodide

Warning

Synonyms: Zinc (II) iodide

223883-50G 223883-250G
220045-192EA 58.77 USD
220045-192 220045-191
Zinc iodide
Zinc iodide
Formula: I₂Zn
MW: 319.2 g/mol
Boiling Pt: 620 °C (1013 hPa)
Melting Pt: 446 °C
Density: 4.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS Number: 10139-47-6
EINECS: 233-396-0
UN: 3260
ADR: 8,II
Merck Index: 12,10269

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR