Potassium hydrogen sulfate, Reagent Grade

Danger

Synonyms: Potassium bisulphate

223476-1KG 223476-500G
220044-857EA 137.23 USD
220044-857 220044-858
Potassium hydrogen sulfate, Reagent Grade
Potassium hydrogen sulphate
Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214 °C
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 13,07695

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR