Dodecamolybdophosphoric acid hydrate ACS

Danger

Synonyms: Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate

221856-25G 221856-5G 221856-100G
220044-548EA 64.62 USD
220044-548 220043-826 220044-547
Dodecamolybdophosphoric acid hydrate ACS
Dodecamolybdophosphoric acid hydrate
Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
MW: 1843.27 g/mol
Melting Pt: 78…90 °C
Density: 1.62 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149913
CAS Number: 51429-74-4
EINECS: 234-713-5
UN: 3260
ADR: 8,III
Merck Index: 12,07498

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR