Copper(II) chloride dihydrate, Reagent Grade, Sigma-Aldrich®

Warning

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

M-Clarity™ quality level = MQ100

221783-500G 221783-100G 221783-2.5KG
220044-538EA 64.62 USD
220044-538 220044-536 220044-537
Copper(II) chloride dihydrate, Reagent Grade, Sigma-Aldrich®
Copper(II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 °C
Density: 2.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR