Mercury(II) iodide ACS
|
Danger
|
Synonyms:
Mercury diiodide, Mercuric iodide
221090-100G
221090-25G
221090-500G
220044-100EA
124.92
USD
220044-100
220044-101
220044-102
Mercury(II) iodide ACS
Mercury(II) iodide
Formula:
HgI₂ MW: 454.4 g/mol Boiling Pt: 354 °C (1013 hPa) Melting Pt: 259 °C Density: 6.36 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011044 CAS Number: 7774-29-0 EINECS: 231-873-8 UN: 1638 ADR: 6.1,II Merck Index: 13,05905 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...