Oxalyl dichloride ≥99%, ReagentPlus®

Danger

Synonyms: Oxalyl chloride

221015-100G 221015-25G 221015-5G
220044-355EA 85.15 USD
220044-355 220044-356 220044-357
Oxalyl dichloride ≥99%, ReagentPlus®
Oxalyl dichloride
Formula: C₂Cl₂O₂
MW: 126.93 g/mol
Boiling Pt: 63 °C (1013 hPa)
Melting Pt: –12 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
UN: 3129
ADR: 4.3,I
Merck Index: 12,07046

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR