Manganese(II) acetate tetrahydrate ≥99%

221007-500G 221007-25G 221007-2KG
220044-354EA 75.08 USD
220044-354 220044-352 220044-353
Manganese(II) acetate tetrahydrate ≥99%
Manganese(II) acetate tetrahydrate
Formula: (H₃CCOO)₂Mn·4H₂O
Melting Pt: 300 °C
Density: 1.58 g/cm³ (20 °C)
MDL Number: MFCD00062552
CAS Number: 6156-78-1
EINECS: 211-334-3
Merck Index: 13,05746

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR