Titanium(III) chloride, ReagentPlus®

Synonyms: Titanium trichloride

220973-25G 220973-5G
220044-091EA 94.15 USD
220044-091 220044-092
Titanium(III) chloride, ReagentPlus®
Titanium(III) chloride
Formula: Cl₃Ti
MW: 154.24 g/mol
Boiling Pt: 960 °C (1013 hPa)
Melting Pt: 440 °C
Density: 1.32 g/cm³ (20 °C)
MDL Number: MFCD00011266
CAS Number: 7705-07-9
EINECS: 231-728-9
UN: 2441
ADR: 4.2,I
Merck Index: 12,09620

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR