Iron(II) chloride tetrahydrate, ReagentPlus®

Danger

Synonyms: Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate

220299-1KG 220299-250G 220299-5G
220044-709EA 148.92 USD
220044-709 220044-710 220044-711
Iron(II) chloride tetrahydrate, ReagentPlus®
Iron(II) chloride tetrahydrate
Formula: FeCl₂·4H₂O
MW: 198.81 g/mol
Melting Pt: 677 °C
Density: 1.926 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149709
CAS Number: 13478-10-9
EINECS: 231-843-4
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR