2,3-Dimethyl-2-butene ≥98%

Synonyms: Tetramethylethylene

220159-100ML 220159-25ML
220044-522EA 674.15 USD
220044-522 220044-523
2,3-Dimethyl-2-butene ≥98%
2,3-Dimethyl-2-butene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 73 °C (1013 hPa)
Melting Pt: -75 °C
Density: 0.7188 g/cm³ (20 °C)
Flash Pt: -16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008897
CAS Number: 563-79-1
EINECS: 209-263-8
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR