L(+)-Potassium sodium tartrate tetrahydrate ≥99% ACS

217255-2.5KG 217255-500G 217255-100G
220043-740EA 224.62 USD
220043-740 220043-741 220043-739
L(+)-Potassium sodium tartrate tetrahydrate ≥99% ACS
L(+)-Potassium sodium tartrate tetrahydrate
Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
Melting Pt: 70…80 °C
Density: ~ 1.767 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
EINECS: 206-156-8
Merck Index: 13,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR