2-Bromo-2-butene cis- and trans-mixture ≥98%

215562-100G 215562-25G
220044-256EA 252 USD
220044-256 220044-257
2-Bromo-2-butene cis- and trans-mixture ≥98%
2-Bromo-2-butene
Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 82 °C (1013 hPa)
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000141
CAS Number: 13294-71-8
EINECS: 236-316-2
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR