Calcium hydride ≥90%, Reagent Grade

Danger

Synonyms: Calcium dihydride

213268-500G 213268-100G
220043-707EA 280 USD
220043-707 220043-706
Calcium hydride ≥90%, Reagent Grade
Calcium hydride
Formula: H₂Ca
MW: 42.09 g/mol
Melting Pt: 816 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010897
CAS Number: 7789-78-8
EINECS: 232-189-2
UN: 1404
ADR: 4.3,I
Merck Index: 13,01674

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR