Terbium(III) chloride hexahydrate ≥99.9%

212903-5G 212903-25G
220043-952EA 137.08 USD
220043-952 220043-951
Terbium(III) chloride hexahydrate ≥99.9%
Terbium(III) chloride hexahydrate
Formula: Cl₃Tb·6H₂O
MW: 373.38 g/mol
Melting Pt: 145 °C
Density: 4.35 g/cm³ (20 °C)
MDL Number: MFCD00149870
CAS Number: 13798-24-8
EINECS: 233-132-4
Merck Index: 12,09300

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR