L(+)-Calcium lactate pentahydrate

Synonyms: L(+)-Lactic acid calcium salt pentahydrate, Calcium (S)-(+)-2-hydroxypropionate pentahydrate, (S)-(+)-2-Hydroxypropionic acid calcium salt pentahydrate, (S)-(+)-Lactic acid calcium salt pentahydrate, (S)-(+)-2-Hydroxypropanoic acid calcium salt pentahydrate, Calcium L(+)-2-hydroxypropionate pentahydrate

21185-250G-F 21185-1KG-F
220043-784EA 42.97 USD
220043-784 220043-783
L(+)-Calcium lactate pentahydrate
L(+)-Calcium lactate pentahydrate
Formula: C₆H₁₀CaO₆·5H₂O
MW: 308.3 g/mol
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00150344
CAS Number: 5743-47-5
EINECS: 248-953-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR