Cesium hydroxide monohydrate

Danger

21000-10G-F 21000-250G-F 21000-U 21000-50G-F
220043-126EA 232.31 USD
220043-126 220043-127 220043-129 220043-128
Cesium hydroxide monohydrate
Cesium hydroxide monohydrate
Formula: HCsO·1H₂O
Melting Pt: 204.85 °C
Density: 3.51 g/cm³ (24.85 °C)
MDL Number: MFCD00149664
CAS Number: 35103-79-8
EINECS: 244-344-1
UN: 3262
ADR: 8,II
Merck Index: 12,02055

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR