Osmium(III) chloride hydrate

209066-500MG 209066-2.5G
220043-501EA 341.54 USD
220043-501 220043-500
Osmium(III) chloride hydrate
Osmium(III) chloride hydrate
Formula: Cl₃Os·H₂O
MDL Number: MFCD00149815
CAS Number: 14996-60-2
EINECS: 236-587-7
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR