Ruthenium(III) chloride

Danger

208523-2G 208523-10G 208523-50G
220042-445EA 82.38 USD
220042-445 220042-444 220042-446
Ruthenium(III) chloride
Ruthenium(III) chloride
Formula: Cl₃Ru
MW: 207.43 g/mol
Melting Pt: 500 °C
Density: 3.11 g/cm³ (20 °C)
MDL Number: MFCD00011208
CAS Number: 10049-08-8
EINECS: 233-167-5
UN: 3260
ADR: 8,III
Merck Index: 12,08455

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR