Calcium iodide hydrate ≥98%

208477-500G 208477-100G
220042-767EA 947.69 USD
220042-767 220043-461
Calcium iodide hydrate ≥98%
Calcium iodide hydrate
Formula: CaI₂·H₂O
MW: 311.9 g/mol
Density: 2.55 g/cm³ (20 °C)
MDL Number: MFCD00149622
CAS Number: 71626-98-7
EINECS: 233-276-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR