Cobalt(II) acetate tetrahydrate, Reagent Grade

Danger

Synonyms: Cobaltous acetate tetrahydrate, Acetic acid cobalt (II) salt tetrahydrate, Cobalt(Ⅱ) acetate tetrahydrate, Cobalt diacetate tetrahydrate

208396-50G 208396-250G 208396-1KG
220043-452EA 39.57 USD
220043-452 220043-451 220043-450
Cobalt(II) acetate tetrahydrate, Reagent Grade
Cobalt(II) acetate tetrahydrate
Formula: (H₃CCOO)₂Co·4H₂O
MW: 249.08 g/mol
Melting Pt: 140 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149569
CAS Number: 6147-53-1
EINECS: 200-755-8
UN: 3077
ADR: 9,III
Merck Index: 12,02494

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR