Silver tetrafluoroborate

Danger

Synonyms: Argentous tetrafluoroborate

208361-50G 208361-1G 208361-10G
220043-447EA 402.92 USD
220043-447 220043-446 220043-445
Silver tetrafluoroborate
Silver tetrafluoroborate
Formula: AgBF₄
MW: 194.67 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00003408
CAS Number: 14104-20-2
EINECS: 237-956-5
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR