Sodium amide ≥90%, Technical Grade

Danger

208329-250G 208329-50G
220042-411EA 72 USD
220042-411 220042-412
Sodium amide ≥90%, Technical Grade
Sodium amide
Formula: H₂NNa
MW: 39.01 g/mol
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 208 °C
Density: 1.39 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011117
CAS Number: 7782-92-5
EINECS: 231-971-0
UN: 1390
ADR: 4.3,II
Merck Index: 12,08717

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR