Thallium(I) sulfate ≥99.9%

Danger

Synonyms: Thallic sulphate, Dithallium sulphate

208191-50G 208191-250G
220043-436EA 303.08 USD
220043-436 220043-435
Thallium(I) sulfate ≥99.9%
Thallium(I) sulphate
Formula: O₄STl₂
MW: 504.83 g/mol
Melting Pt: 632 °C
Density: 6.77 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011278
CAS Number: 7446-18-6
EINECS: 231-201-3
UN: 1707
ADR: 6.1,II
Merck Index: 12,09405

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR