Sodium hexafluorophosphate ≥98%

208051-250G 208051-50G 208051-10G
220043-415EA 1141.54 USD
220043-415 220043-416 220043-414
Sodium hexafluorophosphate ≥98%
Sodium hexafluorophosphate
Formula: F₆NaP
MW: 167.95 g/mol
Melting Pt: 450 °C
Density: 2.505 g/cm³ (20 °C)
MDL Number: MFCD00011122
CAS Number: 21324-39-0
EINECS: 244-333-1
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR