Hydrazinium hydroxide ≥98% N₂H₄ 64-65 %, Reagent Grade, Sigma-Aldrich®

Danger

Synonyms: Hydrazine hydrate, Hydrazine monohydrate

207942-2KG 207942-500G 207942-100G 207942-5G
220043-339EA 299.08 USD
220043-339 220043-340 220043-338 220043-341
Hydrazinium hydroxide ≥98% N₂H₄ 64-65 %, Reagent Grade, Sigma-Aldrich®
Hydrazinium hydroxide
Formula: NH₂NH₂·H₂O
MW: 50.06 g/mol
Boiling Pt: 117.2 °C (1013 hPa)
Melting Pt: -51.7 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00149931
CAS Number: 7803-57-8
EINECS: 206-114-9
UN: 2030
ADR: 8,II
Merck Index: 13,04790

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR