Iridium(III) chloride hydrate, Reagent Grade

Warning

Synonyms: Iridium trichloride hydrate, Iridium(III) chloride xhydrate

206245-5G 206245-1G
220043-235EA 2092.31 USD
220043-235 220043-234
Iridium(III) chloride hydrate, Reagent Grade
Iridium(III) chloride hydrate
Formula: Cl₃Ir·xH₂O
MW: 298.58 g/mol (anhy)
Density: 5.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149750
CAS Number: 14996-61-3
EINECS: 233-044-6
Merck Index: 14,05088

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR