Iridium(IV) oxide

Synonyms: Iridium dioxide, Iridium(IV) oxide

206237-1G 206237-5G
220043-232EA 992.31 USD
220043-232 220043-233
Iridium(IV) oxide
Iridium(IV) oxide
Formula: IrO₂
MW: 224.22 g/mol
Boiling Pt: -27 °C (1013 hPa)
Melting Pt: -102 °C
Density: 3.09 g/cm³ (18 °C)
MDL Number: MFCD00011065
CAS Number: 12030-49-8
EINECS: 234-743-9
UN: 1479
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR