Ruthenium(III) chloride hydrate, ReagentPlus®

Danger

Synonyms: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

206229-5G 206229-25G 206229-1G
220043-231EA 292.31 USD
220043-231 220043-230 220043-229
Ruthenium(III) chloride hydrate, ReagentPlus®
Ruthenium(III) chloride hydrate
Formula: Cl₃Ru·xH₂O
MW: 207.43 g/mol (anhy)
Melting Pt: ? 300 °C
Density: 3.11 g/cm³ (20 °C)
MDL Number: MFCD00149844
CAS Number: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR