Palladium(II) chloride, ReagentPlus®

Warning

Synonyms: Palladium dichloride

205885-25G 205885-1G 205885-5G
220043-106EA 1446.92 USD
220043-106 220043-105 220043-107
Palladium(II) chloride, ReagentPlus®
Palladium(II) chloride
Formula: PdCl₂
MW: 177.33 g/mol
Melting Pt: 678 °C
Density: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,III
Merck Index: 13,07058

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR