Palladium(II) acetate ≥98%, Reagent Grade

Synonyms: Palladium diacetate, Palladium (II) acetate

205869-2G 205869-10G 205869-1G
220043-101EA 135 USD
220043-101 220043-099 220043-100
Palladium(II) acetate ≥98%, Reagent Grade
Palladium(II) acetate
Formula: C₄H₆O₄Pd
MW: 224.51 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR