Zinc oxide, ReagentPlus®

Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

205532-5KG 205532-1KG 205532-100G
220042-995EA 148.92 USD
220042-995 220042-994 220042-993
Zinc oxide, ReagentPlus®
Zinc oxide
Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR