Di-tert-butyl pyrocarbonate ≥99%, ReagentPlus®

Danger

Synonyms: Boc Anhydride, Di-tert-butyl dicarbonate, DIBOC

205249-15KG 205249-50G 205249-1KG 205249-100G 205249-10G
220042-765EA 3923.08 USD
220042-765 220042-481 220042-766 220042-763 220042-764
Di-tert-butyl pyrocarbonate ≥99%, ReagentPlus®
Di-tert-butyl pyrocarbonate
Formula: C₁₀H₁₈O₅
MW: 218.25 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 20…23 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00008805
CAS Number: 24424-99-5
EINECS: 246-240-1
UN: 2930
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae68e9d9dc1f713c2e5ac96bd05040cec5c7bcdd&vid=719e05eb5ef4a6a4176af5e827ff4390d03d64ab&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR