Boron tribromide ≥99.9%, ReagentPlus®

Danger

202207-100G 202207-25G 202207-250G 202207-2KG
220042-581EA 274.62 USD
220042-581 220042-583 220042-582 220042-584
Boron tribromide ≥99.9%, ReagentPlus®
Boron tribromide
Formula: BBr₃
Boiling Pt: 91.2 °C (1013 hPa)
Melting Pt: –46 °C
Density: 2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011312
CAS Number: 10294-33-4
EINECS: 233-657-9
UN: 2692
ADR: 8,I
Merck Index: 13,01337

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR