Cobalt(II) chloride hexahydrate, Reagent Grade

Danger

202185-100G 202185-250G 202185-25G 202185-1KG
220042-572EA 38.22 USD
220042-572 220042-574 220042-575 220042-573
Cobalt(II) chloride hexahydrate, Reagent Grade
Cobalt(II) chloride hexahydrate
Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: 56 °C
Density: 1.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 13,02462

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR